Learn More
Enzo Life Sciences MMP-3 Fluorescent Assay Buffer (20ml)
SDP
Catalog No. 502011090
Encompass_Preferred
Formulation: 50mM MES, 10mM CaCl2, 0.05% Brij-35, pH 6.0. Long Term Storage: -20°C.
Catalog No. 50-201-1090 Supplier Enzo Life Sciences Supplier No. BMLKI1270020
Formulation: 50mM MES, 10mM CaCl2, 0.05% Brij-35, pH 6.0. Long Term Storage: -20°C.