Learn More
Enzo Life Sciences Matrix Metalloproteinase-7 (MMP-7) colorimetric drug discovery kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences BMLAK4060001
Encompass_Preferred
A QUANTIZYME Assay System. The MMP-7 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-7 (MMP-7, matrilysin) using a thiopeptide as a chromogenic substrate The assays are performed in a convenient 96-well microplate format. Included are active enzyme, assay buffer, a prototypic control inhibitor (NNGH), and a detailed instruction booklet. Alternative name: Matrilysin, Pump. Applications: Colorimetric detection, HTS Activity assay.
Catalog No. 50-201-0268