Learn More
Aviva Systems Biology Corp MMP9 Antibody - N-terminal region (ARP33090_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33090_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MMP9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MMP9. Alternate names: GELB, CLG4B, MMP-9, MANDP2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-54879