Learn More
ACROBiosystems Recombinant Protein;Human MMP-9 Protein;HEK293;50UG;MM9,MMP9,CLG4B,GELB,MMP-9,MANDP2,Matrix metallopeptidase 9,Gelatinase B
Catalog No. 501657707
Encompass_Preferred
Recombinant Protein;Human MMP-9 Protein;50UG;Human MMP-9, His Tag (MM9-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Pro 469 (Accession # AAH06093).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 50.8 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7707 Supplier ACROBiosystems Supplier No. MM9H522150UG
Recombinant Protein;Human MMP-9 Protein;50UG;Human MMP-9, His Tag (MM9-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Pro 469 (Accession # AAH06093).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 50.8 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.