Learn More
Medchemexpress LLC Mmpip hydrochloride | 1215566-78-1 | 99.1% | 369.80 g/mol | C19H16ClN3O3 | 1 ML
SDP
Catalog No. 5000245864
Encompass_Preferred
MMPIP hydrochloride is an allosteric metabotropic glutamate receptor 7 (mGluR7) selective antagonist supplied as a ready-to-use 10 mM solution in dimethyl sulfoxide (DMSO). It is intended for in vitro pharmacology and receptor modulation studies and is provided with high purity and characterized identity.
  • Allosteric mGluR7 antagonist for receptor modulation studies.
  • Provided as a 10 mM solution in DMSO for immediate use.
  • High purity suitable for pharmacological assays.
  • Available in multiple solid and solution pack sizes for flexibility.
  • Characterized by CAS 1215566-78-1 and confirmed molecular weight of 369.80 g/mol.
Catalog No. 50-002-45864 Supplier Medchemexpress LLC Supplier No. HY10311110MM 1ML
MMPIP hydrochloride is an allosteric metabotropic glutamate receptor 7 (mGluR7) selective antagonist supplied as a ready-to-use 10 mM solution in dimethyl sulfoxide (DMSO). It is intended for in vitro pharmacology and receptor modulation studies and is provided with high purity and characterized identity.
  • Allosteric mGluR7 antagonist for receptor modulation studies.
  • Provided as a 10 mM solution in DMSO for immediate use.
  • High purity suitable for pharmacological assays.
  • Available in multiple solid and solution pack sizes for flexibility.
  • Characterized by CAS 1215566-78-1 and confirmed molecular weight of 369.80 g/mol.