Learn More
Abcam MMPSI, Caspase-3/7 Inhibitor I, caspase 3 and 7 inhibitor, 5MG
Catalog No. 502705452
Encompass_Preferred
MW 324.35 Da, Purity >98%. Selective caspase 3 and 7 inhibitor (IC₅₀ = 1.7 μM, caspase-3). Reduces ischemic-induced apoptosis (IC₅₀ = 0.5 μM). Shows cardioprotective effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5452 Supplier Abcam Supplier No. AB1450465MG
MW 324.35 Da, Purity >98%. Selective caspase 3 and 7 inhibitor (IC₅₀ = 1.7 μM, caspase-3). Reduces ischemic-induced apoptosis (IC₅₀ = 0.5 μM). Shows cardioprotective effects.
The product is subject to the following: Abcam Restricted Use Statement