Learn More
Maine Molecular Quality Controls Inc ePlex RP Control M306
SDP
Supplier:  Maine Molecular Quality Controls Inc M306
Encompass
The eplex RP Control M306 is intended for use as an external positive and negative quality control for the qualitative detection of pathogens on the cobas eplex respiratory pathogen (RP) panel performed on the cobas eplex system (Roche Diagnostics). The eplex RP Control M306 is composed of synthetic DNA and RNA specifically designed for and intended to be used solely with the cobas eplex RP panel on the cobas eplex system. This product is not intended to replace manufacturer internal eplex RP panel controls provided with the device. eplex RP Control M306 is processed like a patient sample. eplex RP Control M306 is composed of 5 controls, eplex RP Positive A, eplex RP Positive B, eplex RP Positive C, eplex RP Positive D and eplex RP Negative. The eplex RP Control M306 positive controls contain surrogate control material composed of synthetic DNA and RNA corresponding to genome segments of pathogens. eplex RP Negative contains non-target DNA and RNA.
Catalog No. NC1373068