Learn More
Maine Molecular Quality Controls Inc Verigene RP Flex Control Panel M270
SDP
Supplier:  Maine Molecular Quality Controls Inc M270
Encompass
The Verigene RP Flex Control Panel M270 is intended for in vitro use as a quality control to monitor the amplification, detection and identification of multiple viral and bacterial respiratory pathogens as performed by the VERIGENE Respiratory Pathogens Flex Nucleic Acid Test (RP Flex) on the VERIGENE System. Verigene RP Flex Control Panel M270 is composed of 3 controls, Verigene RP Flex Positive A (Positive A), Verigene RP Flex Positive B (Positive B), and Verigene RP Flex Negative. Verigene RP Flex Negative contains non-target RNA. Each Verigene RP Flex Control Panel M270 is composed of 4 tubes of Positive A and 4 tubes of Positive B, 200µL each, containing synthetic target RNA suspended in a non-infectious solution of buffers, preservatives and stabilizers, and 4 tubes of Verigene RP Flex Negative, 200µL each, of synthetic non-target RNA suspended in a non-infectious solution of buffers, preservatives and stabilizers.
Catalog No. NC1050617