Learn More
MOLTOX Molecular Toxicology Inc Methyl Methane-Sulfonate in DMSO, 25 ul/vial, 5 vials/pack, Store in Ultralow
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 60108A25
Encompass
For use in E. coli mutagenicity test with E. coli WP2 tester strains. Dose 2.5 µL/plate. This product comes in neat format. Add organic solvents such as DMSO or sterile dH2O as applicable. Direct acting mutagens do not require the use of S9 metabolic activation to cause mutagenicity. CONTROLCHEM™ chemicals are obtained from major suppliers of research chemicals and are employed without further characterization or purification. Packaged quantities are precise within 1%. Please Note: CONTROLCHEM™ chemicals are known mutagens/carcinogens/toxins and are sold only to those experienced in the safe handling and disposal of hazardous materials. Consult your Institutional Safety Officer before ordering.
Catalog No. 50-948-890