Learn More
Medchemexpress LLC mmu-miR-155-5p mimic 20nmol | 20nmol
SDP
Catalog No. 5000322287
Encompass_Preferred
mmu-miR-155-5p mimics are small chemically synthesized double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity
Catalog No. 50-003-22287 Supplier Medchemexpress LLC Supplier No. HYR0269120NMOL
mmu-miR-155-5p mimics are small chemically synthesized double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity