Learn More
Medchemexpress LLC mmu-miR-378a-3p inhi 20nmol | 20nmol
SDP
Catalog No. 5000322238
Encompass_Preferred
mmu-miR-378a-3p inhibitors are chemically-modified oligonucleotides that hybridize with mature miRNAs The miRNA inhibitors have full-length nucleotide 2 -methoxy modification The miRNA inhibitors strongly compete with mature miRNAs to prevent the complementary pairing of miRNAs and their target genes thereby inhibiting miRNAs from functioning
Catalog No. 50-003-22238 Supplier Medchemexpress LLC Supplier No. HYRI0311320NMOL
mmu-miR-378a-3p inhibitors are chemically-modified oligonucleotides that hybridize with mature miRNAs The miRNA inhibitors have full-length nucleotide 2 -methoxy modification The miRNA inhibitors strongly compete with mature miRNAs to prevent the complementary pairing of miRNAs and their target genes thereby inhibiting miRNAs from functioning