Learn More
Medchemexpress LLC HY-123561 5mg Medchemexpress, MMV008138 CAS:1679333-73-3 Purity:>98%
SDP
Catalog No. 501932581
Encompass_Preferred
Medchemexpress, HY-123561 5mg MMV008138 CAS:1679333-73-3 MMV008138 is a species-selective IspD (enzyme 2-C-methyl-d-erythritol 4-phosphate cytidylyltransferase) -targeting antimalarial agent, with an IC 50 of 44 nM for PfIspD (P. falciparum IspD). MMV008138 inhibits the growth of P. falciparum Dd2 strain with an IC 50 of 250 nM [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2581 Supplier Medchemexpress LLC Supplier No. HY1235615MG
Medchemexpress, HY-123561 5mg MMV008138 CAS:1679333-73-3 MMV008138 is a species-selective IspD (enzyme 2-C-methyl-d-erythritol 4-phosphate cytidylyltransferase) -targeting antimalarial agent, with an IC 50 of 44 nM for PfIspD (P. falciparum IspD). MMV008138 inhibits the growth of P. falciparum Dd2 strain with an IC 50 of 250 nM [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.