Learn More
Cayman Chemical Mn(III) Protoporphyrin IX Chloride, Purity: ≥95%, CAS: 120389-54-0, 500 mg
SDP
Catalog No. 502988250
Encompass_Preferred
A metalloporphyrin; induces mRNA expression of the genes encoding δ-aminolevulinate synthase and HO in chick embryo liver cells at 10 µM; manganese(III) protoporphyrin IX chloride-containing nanobialys have been used in MRI of fibrin clots in vitro; has been used in the synthesis of MnGGH
Catalog No. 50-298-8250 Supplier Cayman Chemical Supplier No. 30808500 MG
A metalloporphyrin; induces mRNA expression of the genes encoding δ-aminolevulinate synthase and HO in chick embryo liver cells at 10 µM; manganese(III) protoporphyrin IX chloride-containing nanobialys have been used in MRI of fibrin clots in vitro; has been used in the synthesis of MnGGH