Learn More
Cayman Chemical Mn IIITBAPchlorIde 10mg
SDP
Catalog No. 502894091
Encompass_Preferred
Mn(III)TBAP is a cell-permeable SOD mimic. At a concentration of 30 µM, it decreases the doubling time of SOD-null E. coli. by 2-fold.{4009}
Catalog No. 50-289-4091 Supplier Cayman Chemical Supplier No. 7585010 MG
Mn(III)TBAP is a cell-permeable SOD mimic. At a concentration of 30 µM, it decreases the doubling time of SOD-null E. coli. by 2-fold.{4009}