Learn More
Cayman Chemical Mn IIITMPyP 10mg
SDP
Catalog No. 502894994
Encompass_Preferred
Mn(III)TMPyP is a cell-permeable SOD mimic and peroxynitrite decomposition catalyst.{6039} The rate constants for superoxide dismutation and peroxynitrite decomposition are 3.9 x 107 M−1s−1 and ~2 x 106 M−1s−1, respectively.{6039,4009}
Catalog No. 50-289-4994 Supplier Cayman Chemical Supplier No. 7585210 MG
Mn(III)TMPyP is a cell-permeable SOD mimic and peroxynitrite decomposition catalyst.{6039} The rate constants for superoxide dismutation and peroxynitrite decomposition are 3.9 x 107 M−1s−1 and ~2 x 106 M−1s−1, respectively.{6039,4009}