Learn More
Sterlitech Corporation Microdyn Nadir, Nadir PM UP020, 20kDa, PES, UF, 297 x 210mm, 1/pk
SDP
Catalog No. 501952334
Encompass_Preferred
Sterlitech ultrafiltration (UF) flat sheet membranes may be used in our process filtration units for evaluation or for any of your general separations needs.
Catalog No. 50-195-2334 Supplier Sterlitech Corporation Supplier No. 1120851
Add to Cart
Add to Cart
Sterlitech ultrafiltration (UF) flat sheet membranes may be used in our process filtration units for evaluation or for any of your general separations needs.