Learn More
Enzo Life Sciences Mn SOD polyclonal antibody (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISOD110F
Encompass_Preferred
Host: Rabbit. Immunogen: Human Mn SOD protein. UniProt: P04179. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Monkey, Hamster, Guinea pig, Dog, Bovine. Applications: IHC, WB. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-412