Learn More
Enzo Life Sciences Mn SOD polyclonal antibody (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISOD111D
Encompass_Preferred
Host: Rabbit. Immunogen: Native rat Mn SOD. UniProt: P07895. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Drosophila, Dog, Clam, Chicken, Bovine. Applications: ELISA. Flow Cytometry, IHC (PS), IP, WB. Recommended dilutions: Flow Cytometry (1:100). Immunoprecipitation (1:100). Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Immunoaffinity purified. Formulation: Liquid. In sodium phosphate buffer, pH 7.0, containing 15mg/ml BSA, 0.09% sodium azide, and 50% glycerol. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-415