Learn More
Enzo Life Sciences Mn SOD (mouse), (recombinant) (His-tag) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISOD200F
Encompass_Preferred
Alternative name: Superoxide Dismutase. SOD2. MW: ~25kDa. Purity: ≥90% (SDS-PAGE; Western blot). Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 1.5 % glycerol and 80mM sodium chloride. Source: Produced in E. coli. Mouse Mn SOD is fused at the N-terminus to a His-tag. UniProt: P09671. Application: WB, Activity assay. Long Term Storage: -80°C
Catalog No. 50-950-081