Learn More
Macherey-Nagel MN SQ WELL BLOCK 4 PC
Catalog No. NC2243114
Encompass_Preferred
96‑well blocks with 2.1 mL square wells.MACHEREY NAGEL INC, mfg# 740476
This item is not returnable. View return policy
Catalog No. NC2243114 Supplier Macherey-Nagel Supplier No. 740476
96‑well blocks with 2.1 mL square wells.MACHEREY NAGEL INC, mfg# 740476