Learn More
Macherey-Nagel MN WASH PLATE
Catalog No. NC3404822
Encompass_Preferred
96‑well plates to facilitate drying of the NucleoSpin 8 / 96 Binding Strips / Plates and to minimize the risk of cross-contamination 4/PK
Catalog No. NC3404822 Supplier Macherey-Nagel Supplier No. 740479
96‑well plates to facilitate drying of the NucleoSpin 8 / 96 Binding Strips / Plates and to minimize the risk of cross-contamination 4/PK