Learn More
Medchemexpress LLC HY-108431 1mg Medchemexpress, MN58b CAS:203192-01-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1084311MG
Encompass_Preferred
Medchemexpress, HY-108431 1mg MN58b CAS:203192-01-2 MN58b is a selective choline kinase α (CHKα) inhibitor, and results in inhibition of phosphocholine synthesis. MN58b reduces cell growth through the induction of apoptosis, and also has antitumoral activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8546