Learn More
Aviva Systems Biology Corp MNDA Antibody - C-terminal region (ARP38152_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38152_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MNDA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MNDA. Alternate names: PYHIN3ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56539