Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci MNDA (CT) Peptide
SDP

Catalog No. 16595360
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-360 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-360 Supplier ProSci Supplier No. 9645P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

MNDA (CT) Peptideis used for blocking the activity of theMNDA (CT) Antibody

MNDA Antibody: MNDA controls the expression of MCL-1 and BCL02 in chronic lymphocytic leukemia cells. MNDA is also a useful marker in the recognition and differential diagnosis of marginal zone lymphomas. MNDA has a role in regulating programmed cell death in myeloid progenitor cells, and its down-regulation in myelodysplastic syndromes is related to granulocyte-macrophage progenitor cell sensitivity to TRAIL-induced programmed cell death.

Specifications

Gene ID (Entrez) 4332
Species Human
Accession Number P41218.1
Common Name myeloid cell nuclear differentiation antigen
Concentration 200 μg/mL
Content And Storage Store MNDA (CT) Peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol MNDA
Quantity 0.05 mg
Sequence 13 amino acids near the carboxy terminus of MNDA (CT).
Synonym MNDA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.