Learn More
Cayman Chemical Mandipropamid, Purity: ≥95%, CAS: 374726-62-2, 50 mg
SDP
Catalog No. 502978398
Encompass_Preferred
A fungicide; inhibits cellulose synthesis and germ tube elongation in P. infestans at 68 nM; reduces mycelial growth of Phytophthora and P. litchi (EC50s = 0.008-0.03 mg/L); toxic to zebrafish embryos, larvae, and adults (LC50s = 19.91, 4.53, and 4.85 mg/L, respectively); has been used as a chemical inducer of proximity to study protein translocation in vitro and in vivo
Catalog No. 50-297-8398 Supplier Cayman Chemical Supplier No. 4012950 MG
A fungicide; inhibits cellulose synthesis and germ tube elongation in P. infestans at 68 nM; reduces mycelial growth of Phytophthora and P. litchi (EC50s = 0.008-0.03 mg/L); toxic to zebrafish embryos, larvae, and adults (LC50s = 19.91, 4.53, and 4.85 mg/L, respectively); has been used as a chemical inducer of proximity to study protein translocation in vitro and in vivo