Learn More
Montana Molecular mNeon Green Fluorescent Protein BacMam, 5mL
SDP
Catalog No. NC2093761
Encompass_Preferred
Express the mNeonGreen fluorescent protein in your cells of interest. The assay is genetically encoded and packaged in BacMam for efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in many primary or iPSC-derived cells. Use in conjunction with Montana Molecular's other BacMam-packaged assays for cAMP, DAG, ER Stress, and Arrestin to optimize transduction conditions in your cells or fluorescence detection settings on your instrument.
Catalog No. NC2093761 Supplier Montana Molecular Supplier No. F0500G005
Express the mNeonGreen fluorescent protein in your cells of interest. The assay is genetically encoded and packaged in BacMam for efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in many primary or iPSC-derived cells. Use in conjunction with Montana Molecular's other BacMam-packaged assays for cAMP, DAG, ER Stress, and Arrestin to optimize transduction conditions in your cells or fluorescence detection settings on your instrument.