Learn More
Proteintech Group Inc mNeonGreen recombinant antibody, VHH-rabbit IgG Fc fusion
SDP
Supplier:  Proteintech Group Inc NFRB20
Encompass
Rabbit monoclonal antibody to mNeonGreen. Recombinant fusion of anti-mNeonGreen VHH [CTK0203] and rabbit IgG Fc domain.
Catalog No. 50-225-8540