Learn More
Proteintech Group Inc mNeonGreen-Trap Agarose beads
SDP
Supplier:  Proteintech Group Inc NTA20
Encompass
mNeonGreen-Trap Agarose is an affinity reagent for IP of mNeonGreen-fusion proteins. It consists of a mNeonGreen Nanobody/ VHH coupled to agarose beads.
Catalog No. 50-225-8544