Learn More
Proteintech Group Inc mNeonGreen-Trap Magnetic Agarose
SDP
Supplier:  Proteintech Group Inc NTMA20
Encompass
The ChromoTek mNeonGreen-Trap Magnetic Agarose is an affinity resin for IP of mNeonGreen-fusion proteins. It consists of a mNeonGreen Nanobody/ VHH coupled to magnetic agarose beads.
Catalog No. 50-225-8551