Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polycarbonate, Four Ports, Manifolds with Female Luer Locks, 180 Rotation, Non-Sterile; 10/PK
Catalog No. NC0173235
Encompass
Polycarbonate manifolds have polyurethane stems and HDPE handles. Ports have female luer locks.
Catalog No. NC0173235 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3060042
Polycarbonate manifolds have polyurethane stems and HDPE handles. Ports have female luer locks.