Learn More
TARGETMOL CHEMICALS INC MNI 137 5MG
SDP
Catalog No. 502580900
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. MNI 137 is a negative allosteric modulator of mGlu2. MNI 137 inhibits glutamate-induced calcium mobilization with IC50s of 8.3 and 12.6 nM for human and rat. purity: 97%
Catalog No. 50-258-0900 Supplier TARGETMOL CHEMICALS INC Supplier No. T230105MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. MNI 137 is a negative allosteric modulator of mGlu2. MNI 137 inhibits glutamate-induced calcium mobilization with IC50s of 8.3 and 12.6 nM for human and rat. purity: 97%