Learn More
Cayman Chemical MNI-caged-L-glutamate, Purity: ≥95%, CAS: 295325-62-1, 25 mg
SDP
Catalog No. 502895648
Encompass_Preferred
A photocleavable derivative of L-glutamate; upon exposure to light, MNI is cleaved and L-glutamate is released; has been used in the study of neural circuits and synaptic regulation of autophagic vacuole motility in dendrites
Catalog No. 50-289-5648 Supplier Cayman Chemical Supplier No. 900455725 MG
A photocleavable derivative of L-glutamate; upon exposure to light, MNI is cleaved and L-glutamate is released; has been used in the study of neural circuits and synaptic regulation of autophagic vacuole motility in dendrites