Learn More
Cayman Chemical MNI-caged-L-glutamate (trifluoroacetate salt), Purity: ≥95%, CAS: 2226362-29-2, 10 mg
SDP
Catalog No. 502984422
Encompass_Preferred
A form of glutamate linked to a photoprotecting that upon exposure to light (300 - 380 nm excitation) releases L-glutamate within submicroseconds; used to investigate the mechanisms of fast synaptic glutamate receptors in situ
Catalog No. 50-298-4422 Supplier Cayman Chemical Supplier No. 1953210 MG
A form of glutamate linked to a photoprotecting that upon exposure to light (300 - 380 nm excitation) releases L-glutamate within submicroseconds; used to investigate the mechanisms of fast synaptic glutamate receptors in situ