Learn More
Carnabio Usa Inc MNK1(MKNK1)/10UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 0214510
Encompass
MNK1(MKNK1), Full-length, Active mutant [T344D], Amino Acid 1-424, BAA19885.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0880