Learn More
Sino Biological Recombinant Rat c-MET/HGFR Protein (GST Tag), Active
Catalog No. 502574202
Encompass_Preferred
Recombinant rat MET (958-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4202 Supplier Sino Biological Supplier No. M5211G500
Recombinant rat MET (958-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.