Learn More
Carnabio Usa Inc MNK2(MKNK2)/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 021465
Encompass
MNK2(MKNK2), Full-length, Active mutant [T379D], Amino Acid 1-465, NP_951009.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0522