Learn More
Innovative Research Inc Mnky Cyno Albumin Pur 25mg
Catalog No. 502744226
Encompass_Preferred
Monkey Cynomolgus Albumin Purified 25mg - Monkey Cynomolgus Albumin Purified from Innovative Research has been purified from US origin cynomolgus (cyno) monkey plasma using proprietary chromatographic and cold ethanol fractionation methods. This is a salt free lyophilized powder with a purity of >95%. This product is useful as a positive control for ELISA and western blot and for a variety of in vitro applications including blocking, cell signaling, drug studies, protein-protein interactions, protein stabilization, receptor binding, and tissue culture. This is an ultra-pure preparation, especially as compared to commercially available heat-shock preparations of Albumin, which are less pure and may contain denatured albumin.
Catalog No. 50-274-4226 Supplier Innovative Research Inc Supplier No. IMNCYALB25MG
Monkey Cynomolgus Albumin Purified 25mg - Monkey Cynomolgus Albumin Purified from Innovative Research has been purified from US origin cynomolgus (cyno) monkey plasma using proprietary chromatographic and cold ethanol fractionation methods. This is a salt free lyophilized powder with a purity of >95%. This product is useful as a positive control for ELISA and western blot and for a variety of in vitro applications including blocking, cell signaling, drug studies, protein-protein interactions, protein stabilization, receptor binding, and tissue culture. This is an ultra-pure preparation, especially as compared to commercially available heat-shock preparations of Albumin, which are less pure and may contain denatured albumin.