Learn More
Innovative Research Inc Mnky Inflam Prt 1Beta Kit
Catalog No. 502744221
Encompass_Preferred
Monkey Macrophage Inflammatory Protein 1 Beta ELISA Kit - This kit is for the quantitative determination of Monkey Macrophage Inflammatory Protein 1 Beta in biofluid samples, such as serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids. This is a sandwich ELISA in strip well format with reagents for up to 96 tests . MIP1b present in samples reacts with an anti-Monkey MIP1b capture antibody coated and dried on a microtiter plate. After appropriate washing steps, an anti-MIP1b detection antibody conjugated to biotin binds to the captured analyte. Excess antibody is washed away and bound antibody is then reacted with avidin conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. Color development is directly proportional to the concentration of active MIP1b in the samples. A standard calibration curve is prepared along with the samples to be measured using dilutions of Monkey MIP1b. For RUO
Catalog No. 50-274-4221 Supplier Innovative Research Inc Supplier No. IMNMIP1BKT
Monkey Macrophage Inflammatory Protein 1 Beta ELISA Kit - This kit is for the quantitative determination of Monkey Macrophage Inflammatory Protein 1 Beta in biofluid samples, such as serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids. This is a sandwich ELISA in strip well format with reagents for up to 96 tests . MIP1b present in samples reacts with an anti-Monkey MIP1b capture antibody coated and dried on a microtiter plate. After appropriate washing steps, an anti-MIP1b detection antibody conjugated to biotin binds to the captured analyte. Excess antibody is washed away and bound antibody is then reacted with avidin conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. Color development is directly proportional to the concentration of active MIP1b in the samples. A standard calibration curve is prepared along with the samples to be measured using dilutions of Monkey MIP1b. For RUO