Learn More
Innovative Research Inc Mnky Intrlkin 1 Rcptr Ant Kit
Supplier:  Innovative Research Inc IMNIL1RAKT
Encompass_Preferred
Monkey Interleukin 1 Receptor Antagonist ELISA Kit - This kit is intended for the quantitative determination of Monkey Interleukin 1 Receptor Antagonist in biofluid samples, such as serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids. This is a sandwich ELISA in strip well format with reagents for up to 96 tests . IL1RA present in samples reacts with an anti-Monkey IL1RA capture antibody coated and dried on a microtiter plate. After appropriate washing steps, an anti-IL1RA detection antibody conjugated to biotin binds to the captured analyte. Excess antibody is washed away and bound antibody is then reacted with avidin conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. Color development is directly proportional to the concentration of active IL1RA in the samples. A standard calibration curve is prepared along with the samples to be measured using dilutions of Monkey IL1RA. For RUO.
Catalog No. 50-274-4222