Learn More
Innovative Research Inc Mnky Plsmngn Act Inhbtr 1 Kit
Catalog No. 502744220
Encompass_Preferred
Monkey Plasminogen Activator Inhibitor 1 ELISA Kit - This kit is for the quantitative determination of Monkey Plasminogen Activator Inhibitor 1 in biofluid samples, such as serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids. This is a sandwich ELISA in strip well format with reagents for up to 96 tests . PAI1 present in samples reacts with an anti-Monkey PAI1 capture antibody coated and dried on a microtiter plate. After appropriate washing steps, an anti-PAI1 detection antibody conjugated to biotin binds to the captured analyte. Excess antibody is washed away and bound antibody is then reacted with avidin conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. Color development is directly proportional to the concentration of active PAI1 in the samples. A standard calibration curve is prepared along with the samples to be measured using dilutions of Monkey PAI1. For research use onlyRUO.
Catalog No. 50-274-4220 Supplier Innovative Research Inc Supplier No. IMNPAI1KT
Monkey Plasminogen Activator Inhibitor 1 ELISA Kit - This kit is for the quantitative determination of Monkey Plasminogen Activator Inhibitor 1 in biofluid samples, such as serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids. This is a sandwich ELISA in strip well format with reagents for up to 96 tests . PAI1 present in samples reacts with an anti-Monkey PAI1 capture antibody coated and dried on a microtiter plate. After appropriate washing steps, an anti-PAI1 detection antibody conjugated to biotin binds to the captured analyte. Excess antibody is washed away and bound antibody is then reacted with avidin conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. Color development is directly proportional to the concentration of active PAI1 in the samples. A standard calibration curve is prepared along with the samples to be measured using dilutions of Monkey PAI1. For research use onlyRUO.