Learn More
New England Biolabs, Inc. MnlI – 2500 units
SDP
Catalog No. 50812068
Encompass
A restriction endonuclease that recognizes the sequence CCTCNNNNNN_N^.
Catalog No. 50-812-068 Supplier New England Biolabs, Inc. Supplier No. R0163L
A restriction endonuclease that recognizes the sequence CCTCNNNNNN_N^.