Learn More
New England Biolabs, Inc. MnlI – 500 units
SDP
Catalog No. 50812069
Encompass
A restriction endonuclease that recognizes the sequence CCTCNNNNNN_N^.
Catalog No. 50-812-069 Supplier New England Biolabs, Inc. Supplier No. R0163S
A restriction endonuclease that recognizes the sequence CCTCNNNNNN_N^.