Learn More
Selleck Chemical LLC MNS (3.4-Methylenedioxy-b-nitrostyrene) S4921-50mg
SDP
Catalog No. 503112095
Encompass_Preferred
MNS is a tyrosine kinase inhibitor inhibits Syk Src p97 with IC50 of 2 5 M 29 3 M and 1 7 M respectively
Catalog No. 50-311-2095 Supplier Selleck Chemical LLC Supplier No. S492150MG
MNS is a tyrosine kinase inhibitor inhibits Syk Src p97 with IC50 of 2 5 M 29 3 M and 1 7 M respectively