Learn More
Apexbio Technology LLC MNS 1485-00-3 200mg
SDP
Catalog No. 5000093922
Encompass_Preferred
MNS (CAS 1485-00-3) is a small molecule selectively inhibiting the tyrosine kinases Src and Syk It demonstrates antiaggregative activity by suppressing GPIIb/IIIa integrin activation with an IC50 of 12 7 M in thrombin-induced platelet aggregation assays MNS does not impact Ca2 -dependent enzymes protein kinase C or arachidonic acid pathways This selective kinase inhibition profile makes MNS a valuable tool for investigating platelet signaling mechanisms and the regulation of integrin-mediated cellular functions
Catalog No. 50-000-93922 Supplier Apexbio Technology LLC Supplier No. A8661200
MNS (CAS 1485-00-3) is a small molecule selectively inhibiting the tyrosine kinases Src and Syk It demonstrates antiaggregative activity by suppressing GPIIb/IIIa integrin activation with an IC50 of 12 7 M in thrombin-induced platelet aggregation assays MNS does not impact Ca2 -dependent enzymes protein kinase C or arachidonic acid pathways This selective kinase inhibition profile makes MNS a valuable tool for investigating platelet signaling mechanisms and the regulation of integrin-mediated cellular functions