Learn More
Biotium Mix-n-Stain™ Biotin Antibody Labeling Kit, 1x(5-20ug) labeling
Supplier:  Biotium 92286
Encompass_Preferred
Mix-n-Stain™ Biotin Antibody Labeling Kits are a revolutionary antibody labeling technology that allows you to label your antibody with biotin in 30 minutes without a purification step. The labeling procedure tolerates many common buffer components and antibody stabilizers. Choose from kit sizes to label 5-20 ug, 20-50 ug, or 50-100 ug of your antibody.
Catalog No. 50-196-4961