Learn More
Biotium Mix-n-Stain™ CF405M Antibody Labeling Kit, 1x(20-50ug) labeling
Catalog No. 501964942
Encompass_Preferred
Mix-n-Stain™ Antibody Labeling Kits are a revolutionary antibody labeling technology that allows you to label your antibody with one of our superior CF dyes in 30 minutes without a purification step. The labeling procedure tolerates many common buffer components and antibody stabilizers. Choose from kit sizes to label 5-20 ug, 20-50 ug, or 50-100 ug of your antibody. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Blue fluorescent CF405M dye has excitation/emission at 408/452 nm. Note: Conjugates of blue fluorescent dyes like CF350, CF405S, and CF405M are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.
Catalog No. 50-196-4942 Supplier Biotium Supplier No. 92252
Mix-n-Stain™ Antibody Labeling Kits are a revolutionary antibody labeling technology that allows you to label your antibody with one of our superior CF dyes in 30 minutes without a purification step. The labeling procedure tolerates many common buffer components and antibody stabilizers. Choose from kit sizes to label 5-20 ug, 20-50 ug, or 50-100 ug of your antibody. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Blue fluorescent CF405M dye has excitation/emission at 408/452 nm. Note: Conjugates of blue fluorescent dyes like CF350, CF405S, and CF405M are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.