Learn More
Biotium Mix-n-Stain™ CF514 Antibody Labeling Kit, 1x(5-20ug) labeling
Supplier:  Biotium 92331
Encompass_Preferred
Mix-n-Stain™ Antibody Labeling Kits are a revolutionary antibody labeling technology that allows you to label your antibody with one of our superior CF dyes in 30 minutes without a purification step. The labeling procedure tolerates many common buffer components and antibody stabilizers. Choose from kit sizes to label 5-20 ug, 20-50 ug, or 50-100 ug of your antibody. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Green fluorescent CF514 dye has excitation/emission at 516/548 nm.
Catalog No. 50-196-4979