Learn More
Biotium Mix-n-Stain™ CF540 Small Ligand Labeling Kit
Catalog No. 501964984
Encompass_Preferred
Mix-n-Stain™ Small Ligand Labeling Kits are designed for rapid labeling of low molecular weight (MW 150 -5000) ligands without a purification step. Suitable ligands or substrates include SNAP-tag, CLIP-tag™ and HaloTag ligand derivatives that have an aliphatic amine. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Orange fluorescent CF540 has excitation/emission at 540/570 nm. It is suitable for labeling ligands for intracellular staining.
Catalog No. 50-196-4984 Supplier Biotium Supplier No. 92358
Mix-n-Stain™ Small Ligand Labeling Kits are designed for rapid labeling of low molecular weight (MW 150 -5000) ligands without a purification step. Suitable ligands or substrates include SNAP-tag, CLIP-tag™ and HaloTag ligand derivatives that have an aliphatic amine. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Orange fluorescent CF540 has excitation/emission at 540/570 nm. It is suitable for labeling ligands for intracellular staining.