Learn More
Biotium Mix-n-Stain™ CF660C Small Ligand Labeling Kit
Catalog No. 501964985
Encompass_Preferred
Mix-n-Stain™ Small Ligand Labeling Kits are designed for rapid labeling of low molecular weight (MW 150 -5000) ligands without a purification step. Suitable ligands or substrates include SNAP-tag, CLIP-tag™ and HaloTag ligand derivatives that have an aliphatic amine. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF660C dye has excitation/emission at 667/685 nm. It is suitable for labeling ligands for cell surface staining.
Catalog No. 50-196-4985 Supplier Biotium Supplier No. 92360
Mix-n-Stain™ Small Ligand Labeling Kits are designed for rapid labeling of low molecular weight (MW 150 -5000) ligands without a purification step. Suitable ligands or substrates include SNAP-tag, CLIP-tag™ and HaloTag ligand derivatives that have an aliphatic amine. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF660C dye has excitation/emission at 667/685 nm. It is suitable for labeling ligands for cell surface staining.