Learn More
Medchemexpress LLC MNS (NSC 170724), 5-(2-nitrovinyl)benzodioxole | 1485-00-3 | 99.55% | C9H7NO4 | 200 MG
SDP
Catalog No. 5000432614
Encompass_Preferred
MNS (NSC 170724), a beta-nitrostyrene derivative, is an orally active tyrosine kinase inhibitor and a broad-spectrum antiplatelet agent. It inhibits Src, Syk, and FAK, and is cytotoxic to various cell types. This compound also inhibits NLRP3 inflammasome and β1 integrin.
  • Acts as a tyrosine kinase inhibitor.
  • Functions as a broad-spectrum antiplatelet agent.
  • Inhibits Src, Syk, and FAK.
  • Inhibits NLRP3 inflammasome.
  • Inhibits β1 integrin.
  • Exhibits cytotoxicity to various cell types.
  • Completely inhibits U46619, ADP-, arachidonic acid-, collagen-, and thrombin-induced platelet aggregation.
Catalog No. 50-004-32614 Supplier Medchemexpress LLC Supplier No. HY78263200MG
MNS (NSC 170724), a beta-nitrostyrene derivative, is an orally active tyrosine kinase inhibitor and a broad-spectrum antiplatelet agent. It inhibits Src, Syk, and FAK, and is cytotoxic to various cell types. This compound also inhibits NLRP3 inflammasome and β1 integrin.
  • Acts as a tyrosine kinase inhibitor.
  • Functions as a broad-spectrum antiplatelet agent.
  • Inhibits Src, Syk, and FAK.
  • Inhibits NLRP3 inflammasome.
  • Inhibits β1 integrin.
  • Exhibits cytotoxicity to various cell types.
  • Completely inhibits U46619, ADP-, arachidonic acid-, collagen-, and thrombin-induced platelet aggregation.